Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNNM2
Entrez Gene ID
504632
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 2
Gene Aliases
-
Chromosome Location
Chr.26: 23550662 - 23734363 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191172.1 NP_001178101.1 2 - 3 1761 116

View Full Product Details