Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTTNBP2NL
Entrez Gene ID
538981
Gene Name
CTTNBP2 N-terminal like
Gene Aliases
-
Chromosome Location
Chr.3: 30833767 - 30886450 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191181.2 NP_001178110.1 2 - 3 237 109
GenBank mRNA BC111189.1 - 5 - 6 402 109

View Full Product Details