Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CEP131
Entrez Gene ID
514944
Gene Name
centrosomal protein 131
Gene Aliases
AZI1
Chromosome Location
Chr.19: 51500742 - 51517016 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191187.3 NP_001178116.3 11 - 12 1457 73

View Full Product Details