Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNMB3
Entrez Gene ID
539609
Gene Name
potassium calcium-activated channel subfamily M regulatory beta subunit 3
Gene Aliases
-
Chromosome Location
Chr.1: 87793394 - 87891502 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191284.2 NP_001178213.1 1 - 2 59 79

View Full Product Details