Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C18H19orf33
Entrez Gene ID
787634
Gene Name
chromosome 18 C19orf33 homolog
Gene Aliases
-
Chromosome Location
Chr.18: 48118686 - 48119892 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001207046.1 NP_001193975.1 1 - 2 130 79
GenBank mRNA BC102086.1 - 1 - 2 131 79
EV616576.1 - 1 - 2 81 79

View Full Product Details