Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNF19A
Entrez Gene ID
540195
Gene Name
ring finger protein 19A, RBR E3 ubiquitin protein ligase
Gene Aliases
-
Chromosome Location
Chr.14: 64047881 - 64103385 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191331.1 NP_001178260.1 9 - 10 2169 98
GenBank mRNA BC151661.1 - 9 - 10 2148 98

View Full Product Details