Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HOXB2
Entrez Gene ID
616129
Gene Name
homeobox B2
Gene Aliases
-
Chromosome Location
Chr.19: 37946400 - 37948660 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191335.2 NP_001178264.1 1 - 2 385 115

View Full Product Details