Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BCL6B
Entrez Gene ID
100138731
Gene Name
BCL6B transcription repressor
Gene Aliases
-
Chromosome Location
Chr.19: 26813286 - 26819327 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191366.2 NP_001178295.2 8 - 9 1368 66

View Full Product Details