Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYNM
Entrez Gene ID
514186
Gene Name
synemin
Gene Aliases
DMN
Chromosome Location
Chr.21: 7579409 - 7606604 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191453.1 NP_001178382.1 3 - 4 999 118

View Full Product Details