Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYLC1
Entrez Gene ID
281737
Gene Name
cylicin 1
Gene Aliases
-
Chromosome Location
Chr.X: 68899563 - 68943644 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174302.3 NP_776727.1 4 - 5 252 98
GenBank mRNA Z22779.1 - 4 - 5 253 98

View Full Product Details