Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EHD4
Entrez Gene ID
505206
Gene Name
EH domain containing 4
Gene Aliases
-
Chromosome Location
Chr.10: 37295608 - 37379520 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192053.1 NP_001178982.1 5 - 6 1186 70

View Full Product Details