Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATG14
Entrez Gene ID
538831
Gene Name
autophagy related 14
Gene Aliases
-
Chromosome Location
Chr.10: 67842143 - 67879035 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192099.1 NP_001179028.1 4 - 5 412 123

View Full Product Details