Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TLX2
Entrez Gene ID
539041
Gene Name
T cell leukemia homeobox 2
Gene Aliases
-
Chromosome Location
Chr.11: 10122554 - 10123956 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192158.1 NP_001179087.1 1 - 2 404 85

View Full Product Details