Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC16A14
Entrez Gene ID
539153
Gene Name
solute carrier family 16 member 14
Gene Aliases
MCT14
Chromosome Location
Chr.2: 118068142 - 118107681 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192187.1 NP_001179116.1 3 - 4 1386 106

View Full Product Details