Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBX4
Entrez Gene ID
508750
Gene Name
T-box transcription factor 4
Gene Aliases
-
Chromosome Location
Chr.19: 11605886 - 11637853 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192193.1 NP_001179122.1 4 - 5 574 70

View Full Product Details