Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALKBH6
Entrez Gene ID
539257
Gene Name
alkB homolog 6
Gene Aliases
-
Chromosome Location
Chr.18: 46681498 - 46687255 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192208.2 NP_001179137.1 3 - 4 201 80

View Full Product Details