Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THUMPD2
Entrez Gene ID
509460
Gene Name
THUMP domain containing 2
Gene Aliases
-
Chromosome Location
Chr.11: 22040726 - 22080810 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192222.3 NP_001179151.2 1 - 2 134 92
GenBank mRNA BC149576.1 - 1 - 2 150 92

View Full Product Details