Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RLIM
Entrez Gene ID
539871
Gene Name
ring finger protein, LIM domain interacting
Gene Aliases
RNF12
Chromosome Location
Chr.X: 76498578 - 76522745 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192322.1 NP_001179251.1 2 - 3 267 103

View Full Product Details