Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC45A1
Entrez Gene ID
513008
Gene Name
solute carrier family 45 member 1
Gene Aliases
-
Chromosome Location
Chr.16: 45035441 - 45057607 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192339.1 NP_001179268.1 8 - 9 2109 83

View Full Product Details