Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RCC1L
Entrez Gene ID
513273
Gene Name
RCC1 like
Gene Aliases
-
Chromosome Location
Chr.25: 32669753 - 32693554 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192346.1 NP_001179275.1 4 - 5 684 67

View Full Product Details