Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SMPD3
Entrez Gene ID
514201
Gene Name
sphingomyelin phosphodiesterase 3
Gene Aliases
NSMase2
Chromosome Location
Chr.18: 35721610 - 35805773 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192363.2 NP_001179292.1 7 - 8 2216 95

View Full Product Details