Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAX9
Entrez Gene ID
540196
Gene Name
paired box 9
Gene Aliases
-
Chromosome Location
Chr.21: 46857069 - 46878787 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192369.1 NP_001179298.1 1 - 2 186 59

View Full Product Details