Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADAMTS15
Entrez Gene ID
514927
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 15
Gene Aliases
-
Chromosome Location
Chr.29: 36560407 - 36582521 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192390.2 NP_001179319.1 3 - 4 1254 111

View Full Product Details