Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRPL10
Entrez Gene ID
515014
Gene Name
mitochondrial ribosomal protein L10
Gene Aliases
L10mt, MRP-L10
Chromosome Location
Chr.19: 38631957 - 38637781 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192395.1 NP_001179324.1 1 - 2 76 82

View Full Product Details