Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARHGAP35
Entrez Gene ID
540310
Gene Name
Rho GTPase activating protein 35
Gene Aliases
GRLF1
Chromosome Location
Chr.18: 53947317 - 54063061 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192398.1 NP_001179327.1 5 - 6 4143 109

View Full Product Details