Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MOB1B
Entrez Gene ID
515407
Gene Name
MOB kinase activator 1B
Gene Aliases
MOBKL1A
Chromosome Location
Chr.6: 86250301 - 86308361 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192412.3 NP_001179341.1 3 - 4 416 66

View Full Product Details