Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAPN14
Entrez Gene ID
515741
Gene Name
calpain 14
Gene Aliases
-
Chromosome Location
Chr.11: 68706369 - 68744134 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192425.1 NP_001179354.1 18 - 19 1900 59

View Full Product Details