Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYT12
Entrez Gene ID
536820
Gene Name
synaptotagmin 12
Gene Aliases
-
Chromosome Location
Chr.29: 45010199 - 45031375 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192890.2 NP_001179819.1 4 - 5 1165 80

View Full Product Details