Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASIC1
Entrez Gene ID
538244
Gene Name
acid sensing ion channel subunit 1
Gene Aliases
ACCN2
Chromosome Location
Chr.5: 29811101 - 29836206 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192933.1 NP_001179862.1 9 - 10 1370 62

View Full Product Details