Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBXN2A
Entrez Gene ID
613578
Gene Name
UBX domain protein 2A
Gene Aliases
UBXD4
Chromosome Location
Chr.11: 75050371 - 75081454 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192967.1 NP_001179896.1 1 - 2 363 89

View Full Product Details