Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBXN2A
Entrez Gene ID
613578
Gene Name
UBX domain protein 2A
Gene Aliases
UBXD4
Chromosome Location
Chr.11: 75050371 - 75081454 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192967.1 NP_001179896.1 7 - 8 1135 92

View Full Product Details