Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNH5
Entrez Gene ID
537813
Gene Name
potassium voltage-gated channel subfamily H member 5
Gene Aliases
-
Chromosome Location
Chr.10: 74940464 - 75324314 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192989.1 NP_001179918.1 3 - 4 300 95

View Full Product Details