Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CERS6
Entrez Gene ID
616901
Gene Name
ceramide synthase 6
Gene Aliases
LASS6
Chromosome Location
Chr.2: 27353327 - 27712377 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193132.1 NP_001180061.1 5 - 6 514 92

View Full Product Details