Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NKX3-2
Entrez Gene ID
616907
Gene Name
NK3 homeobox 2
Gene Aliases
-
Chromosome Location
Chr.6: 108754570 - 108757070 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193133.1 NP_001180062.1 1 - 2 470 71

View Full Product Details