Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MXD3
Entrez Gene ID
533359
Gene Name
MAX dimerization protein 3
Gene Aliases
-
Chromosome Location
Chr.7: 38779721 - 38785698 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193259.1 NP_001180188.1 3 - 4 265 61

View Full Product Details