Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
ANP32A
Entrez Gene ID
538427
Gene Name
acidic nuclear phosphoprotein 32 family member A
Gene Aliases
ANP32D, HPPCn, LANP, PHAPI
Chromosome Location
Chr.10: 15613398 - 15652308 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195019.1 NP_001181948.1 1 - 2 199 78

View Full Product Details