Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARPC1B
Entrez Gene ID
326600
Gene Name
actin related protein 2/3 complex subunit 1B
Gene Aliases
-
Chromosome Location
Chr.25: 37005090 - 37019132 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014844.3 NP_001014844.1 6 - 7 768 60
GenBank mRNA AB099030.1 - 3 - 4 499 60
BC102942.1 - 6 - 7 769 60
BT021526.1 - 5 - 6 721 60
BT021895.1 - 5 - 6 715 60
BT025344.1 - 5 - 6 709 60
BT026160.1 - 5 - 6 710 60
BT026274.1 - 5 - 6 709 60

View Full Product Details