Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
CD81
Entrez Gene ID
511435
Gene Name
CD81 molecule
Gene Aliases
-
Chromosome Location
Chr.29: 49197099 - 49216812 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035099.1 NP_001030271.1 7 - 8 876 62
GenBank mRNA BC102513.1 - 7 - 8 876 62

View Full Product Details