Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THUMPD1
Entrez Gene ID
524854
Gene Name
THUMP domain containing 1
Gene Aliases
-
Chromosome Location
Chr.25: 18449219 - 18456899 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046277.1 NP_001039742.1 1 - 2 246 63
GenBank mRNA BC114098.1 - 1 - 2 246 63

View Full Product Details