Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PALM2
Entrez Gene ID
100532750
Gene Name
paralemmin 2
Gene Aliases
AKAP2, PALM2-AKAP2, PALM2AKAP2
Chromosome Location
Chr.8: 99342276 - 99503025 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204119.1 NP_001191048.1 3 - 4 258 86
NM_001371509.1 NP_001358438.1 3 - 4 482 86
GenBank mRNA DN283780.1 - 2 - 3 181 86
DN511821.1 - 2 - 3 181 86
DN546048.1 - 2 - 3 181 86
EH372970.1 - 3 - 4 272 86

View Full Product Details