Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPR75
Entrez Gene ID
539470
Gene Name
G protein-coupled receptor 75
Gene Aliases
-
Chromosome Location
Chr.11: 36560294 - 36570055 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205061.1 NP_001191990.1 1 - 2 206 87

View Full Product Details