Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DOK3
Entrez Gene ID
514894
Gene Name
docking protein 3
Gene Aliases
-
Chromosome Location
Chr.7: 38998538 - 39010388 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205324.1 NP_001192253.1 1 - 2 124 62

View Full Product Details