Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL42
Entrez Gene ID
781458
Gene Name
kelch like family member 42
Gene Aliases
KLHDC5
Chromosome Location
Chr.5: 82042286 - 82063595 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205415.2 NP_001192344.1 1 - 2 990 59

View Full Product Details