Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
B4GAT1
Entrez Gene ID
618055
Gene Name
beta-1,4-glucuronyltransferase 1
Gene Aliases
B3GNT1
Chromosome Location
Chr.29: 44397096 - 44399327 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034808.1 NP_001029980.1 2 - 2 1716 147
GenBank mRNA BC105206.1 - 2 - 2 1736 147
BT020768.1 - 2 - 2 1716 147
BT021635.1 - 2 - 2 1691 147

View Full Product Details