Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCER1
Entrez Gene ID
615630
Gene Name
coiled-coil glutamate rich protein 1
Gene Aliases
C12orf12, C5H12orf12
Chromosome Location
Chr.5: 20831993 - 20833802 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038578.2 NP_001033667.1 1 - 1 1616 99
GenBank mRNA BC111324.1 - 1 - 1 1617 99

View Full Product Details