Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- XIST
- Entrez Gene ID
-
338325
- Gene Name
- X (inactive)-specific transcript
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 77161577 - 77198299 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
DY130193.1 |
- |
2 - 3 |
114 |
81 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA mir-421 |
MIR421 |
bta-mir-421, mir-421 |
-257.8kb
|
Sense |
Chr. - 76925575 - 76925659 on Build ARS-UCD1.2 |
Downstream |
cysteine rich hydrophobic domain 1 |
CHIC1 |
|
127.2kb
|
Anti-sense |
Chr. - 77312352 - 77359558 on Build ARS-UCD1.2 |
View Full Product Details