Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- H19
- Entrez Gene ID
-
100126192
- Gene Name
- H19, imprinted maternally expressed transcript (non-protein coding)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.29: 49510998 - 49513370 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_003958.2 |
|
1 - 2 |
1116 |
91 |
GenBank mRNA |
DV928713.1 |
- |
1 - 2 |
536 |
91 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
insulin like growth factor 2 |
IGF2 |
|
-88.5kb
|
Sense |
Chr. - 49395153 - 49422469 on Build ARS-UCD1.2 |
Downstream |
mitochondrial ribosomal protein L23 |
MRPL23 |
|
30.4kb
|
Anti-sense |
Chr. - 49542745 - 49549286 on Build ARS-UCD1.2 |
View Full Product Details