Product Details

SNP ID
rs75538511
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241811354 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGGTCCTGACGTAGCCCTGAGA[C/T]GTGGCCAGCTCCCTGGCCTGCTGCC
Phenotype
MIM: 608237 MIM: 608527
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GAL3ST2 PubMed Links

Gene Details

Gene
GAL3ST2
Gene Name
galactose-3-O-sulfotransferase 2
There are no transcripts associated with this gene.

Gene
LOC107986004
Gene Name
uncharacterized LOC107986004
There are no transcripts associated with this gene.

Gene
NEU4
Gene Name
neuraminidase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167599.2 423 UTR 5 NP_001161071.1
NM_001167600.2 423 Intron NP_001161072.1
NM_001167601.2 423 Intron NP_001161073.1
NM_001167602.2 423 UTR 5 NP_001161074.1
NM_080741.3 423 UTR 5 NP_542779.2

View Full Product Details