Product Details

SNP ID
rs79042392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:114065293 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTTACCAAGGTAGTTAATGGCTG[G/T]CGCCTAGGAAAAATAAAAAACCTGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC191 PubMed Links

Gene Details

Gene
CCDC191
Gene Name
coiled-coil domain containing 191
There are no transcripts associated with this gene.

Gene
QTRT2
Gene Name
queuine tRNA-ribosyltransferase accessory subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256835.1 158 Missense Mutation TGG,TGT W24C NP_001243764.1
NM_001256836.1 158 Intron NP_001243765.1
NM_001256837.1 158 Intron NP_001243766.1
NM_024638.3 158 Missense Mutation TGG,TGT W12C NP_078914.1
XM_005247769.3 158 Intron XP_005247826.1
XM_005247770.3 158 UTR 5 XP_005247827.1
XM_006713744.3 158 Intron XP_006713807.1
XM_011513137.2 158 Missense Mutation TGG,TGT W24C XP_011511439.1
XM_017007182.1 158 UTR 5 XP_016862671.1
XM_017007183.1 158 Intron XP_016862672.1
XM_017007184.1 158 UTR 5 XP_016862673.1

View Full Product Details