Product Details

SNP ID
rs78952413
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:134487574 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCATTGCTTCCTGCTTTCAAAAA[A/G]TGTCAACTTGACTTGAAATTCAGCT
Phenotype
MIM: 614484 MIM: 614724
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANAPC13 PubMed Links

Gene Details

Gene
ANAPC13
Gene Name
anaphase promoting complex subunit 13
There are no transcripts associated with this gene.

Gene
CEP63
Gene Name
centrosomal protein 63
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042383.1 Intron NP_001035842.1
NM_001042384.1 Intron NP_001035843.1
NM_001042400.1 Intron NP_001035859.1
NM_025180.3 Intron NP_079456.2
XM_005247795.4 Intron XP_005247852.1
XM_005247797.3 Intron XP_005247854.1
XM_005247799.3 Intron XP_005247856.1
XM_006713760.3 Intron XP_006713823.1
XM_011513194.2 Intron XP_011511496.1
XM_011513197.2 Intron XP_011511499.1
XM_017007244.1 Intron XP_016862733.1
XM_017007245.1 Intron XP_016862734.1
XM_017007246.1 Intron XP_016862735.1
XM_017007247.1 Intron XP_016862736.1
XM_017007248.1 Intron XP_016862737.1
XM_017007249.1 Intron XP_016862738.1
XM_017007250.1 Intron XP_016862739.1
XM_017007251.1 Intron XP_016862740.1
XM_017007252.1 Intron XP_016862741.1
XM_017007253.1 Intron XP_016862742.1
XM_017007254.1 Intron XP_016862743.1
XM_017007255.1 Intron XP_016862744.1
XM_017007256.1 Intron XP_016862745.1
XM_017007257.1 Intron XP_016862746.1
XM_017007258.1 Intron XP_016862747.1
XM_017007259.1 Intron XP_016862748.1
XM_017007260.1 Intron XP_016862749.1
XM_017007261.1 Intron XP_016862750.1
XM_017007262.1 Intron XP_016862751.1
XM_017007263.1 Intron XP_016862752.1
XM_017007264.1 Intron XP_016862753.1
XM_017007265.1 Intron XP_016862754.1
XM_017007266.1 Intron XP_016862755.1
XM_017007267.1 Intron XP_016862756.1
XM_017007268.1 Intron XP_016862757.1
XM_017007269.1 Intron XP_016862758.1

View Full Product Details